Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-Dimethoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M22265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22265G Supplier TCI America Supplier No. M22265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2150-41-6
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00070649
InChI Key IHIJFZWLGPEYPJ-UHFFFAOYSA-N
PubChem CID 137451
IUPAC Name methyl 2,4-dimethoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C=C1OC

Specifications

Color Yellow
Boiling Point 296°C
Quantity 5 g
Synonym 2,4-dimethoxybenzoic acid methyl ester, benzoic acid, 2,4-dimethoxy-, methyl ester, acmc-1ccii, methyl2,4-dimethoxybenzoate, methyl 2,4-dimethoxybenzoate, 2,4-dimethoxy-benzoic acid methyl ester, benzoic acid,2,4-dimethoxy-,methyl ester, benzoic acid,2,4-dimethoxy-, methyl ester, 2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,4-Dimethoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.