Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,5-Dibromobenzoate 98.0+%, TCI America™
SDP

Catalog No. D267325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D267325G 25 g
D26735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D267325G Supplier TCI America Supplier No. D267325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57381-43-8
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.942
MDL Number MFCD00144757
InChI Key RBCUIRIGTNHLPS-UHFFFAOYSA-N
Synonym 2,5-dibromobenzoic acid methyl ester, benzoic acid, 2,5-dibromo-, methyl ester, pubchem3972, acmc-1aybl, #, methyl 2,5-bis bromanyl benzoate, methyl 2,5-dibromobenzoate, benzoic acid,2,5-dibromo-, methyl ester, 3,5-diamino-2-methyl benzoic acid
PubChem CID 554523
IUPAC Name methyl 2,5-dibromobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Br)Br

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 25 g
Formula Weight 293.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,5-Dibromobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.