Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,5-difluoro-4-nitrobenzoate, 95%

Catalog No. AAH63472MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63472MD 250 mg
AAH6347203 1 g
AAH6347206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63472MD Supplier Thermo Scientific Chemicals Supplier No. H63472MD

CAS: 924868-81-5 | C8H5F2NO4 | 217.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924868-81-5
Molecular Formula C8H5F2NO4
Molecular Weight (g/mol) 217.13
MDL Number MFCD08689670
InChI Key XBUVRWIIEREYFL-UHFFFAOYSA-N
Synonym methyl 2,5-difluoro-4-nitrobenzenecarboxylate, methyl2,5-difluoro-4-nitrobenzoate, benzoic acid, 2,5-difluoro-4-nitro-, methyl ester, methyldifluoronitrobenzenecarboxylate, methyl-2,5-difluoro-4-nitrobenzoate, 2,5-difluoro-4-methoxycarbonyl nitrobenzene, 2,5-difluoro-4-nitro-benzoic acid methyl ester
PubChem CID 18526058
IUPAC Name methyl 2,5-difluoro-4-nitrobenzoate
SMILES COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O

Specifications

Quantity 250 mg
Formula Weight 217.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 2,5-difluoro-4-nitrobenzoate
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.