Learn More
Medchemexpress LLC Methyl 2-(6-bromo-1H-indol-3-yl)acetate | 152213-63-3 | MFCD18452338 | 97.4% | 268.11 g/mol | C11H10BrNO2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW035305
Encompass_Preferred
Methyl 2-(6-bromo-1H-indol-3-yl)acetate is a brominated indole-derived small-molecule research reagent, the methyl ester of 6-bromo-indole-3-acetic acid, originally reported from marine sources. It is supplied as a light brown solid (CAS 152213-63-3) with defined analytical characterization for laboratory use.
  • High purity for analytical and synthetic applications.
  • Brominated indole scaffold useful for marine natural product studies and medicinal chemistry.
  • Well-characterized molecular properties: C11H10BrNO2, 268.11 g/mol.
  • Stable solid with room-temperature shelf life and recommended low-temperature solvent storage.
  • Supplied with a certificate of analysis for lot-specific verification.
Catalog No. 50-003-03060