Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Methyl 2,6-dichlorobenzoate, 97%

Catalog No. AAB2491203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2491203 1 g
AAB2491206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2491203 Supplier Thermo Scientific™ Supplier No. B2491203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 14920-87-7 | C8H6Cl2O2 | 205.03 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14920-87-7
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD01313701
InChI Key HEYGSGDIWJDORA-UHFFFAOYSA-N
Synonym 2,6-Dichlorobenzoic acid methyl ester;
IUPAC Name methyl 2,6-dichlorobenzoate
SMILES COC(=O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 25-30°
Boiling Point 250°
Quantity 1 g
Formula Weight 205.04
Physical Form Solid
ShelfLife 5 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.