Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-amino-5-(trifluoromethyl)benzoate, 98%

Catalog No. AAH6428306
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH6428306

Supplier: Thermo Scientific Chemicals H6428306

Only null left
Add to Cart

CAS: 117324-58-0 | C9H8F3NO2 | 219.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117324-58-0
Molecular Formula C9H8F3NO2
Molecular Weight (g/mol) 219.163
MDL Number MFCD08234902
InChI Key QGFUDNJZHZNPCS-UHFFFAOYSA-N
PubChem CID 14233808
IUPAC Name methyl 2-amino-5-(trifluoromethyl)benzoate
SMILES COC(=O)C1=C(C=CC(=C1)C(F)(F)F)N

Specifications

Color White
Quantity 5 g
Synonym methyl 2-amino-5-trifluoromethyl benzoate, methyl2-amino-5-trifluoromethyl benzoate, 2-amino-5-trifluoromethyl benzoic acid methyl ester, benzoic acid,2-amino-5-trifluoromethyl-, methyl ester, benzoic acid, 2-amino-5-trifluoromethyl-, methyl ester, acmc-209z2m, benzoicacid,2-amino-5-trifluoromethyl-,methylester, methyl 5-trifluoromethylanthranilate, 2-amino-5-trifluoromethyl-benzoic acid methyl ester
Formula Weight 219.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-5-(trifluoromethyl)benzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.