Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromo-3-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. M254625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M254625G 25 g
M25465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M254625G Supplier TCI America Supplier No. M254625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131001-86-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00144762
InChI Key QAOFGUXVDAZKBW-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
PubChem CID 10823279
IUPAC Name methyl 2-bromo-3-methylbenzoate
SMILES COC(=O)C1=C(Br)C(C)=CC=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromo-3-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.