Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-bromo-4-methylbenzoate, 98%

Catalog No. AAH63144MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63144MD 250 mg
AAH6314403 1 g
AAH6314406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63144MD Supplier Thermo Scientific Chemicals Supplier No. H63144MD

CAS: 87808-49-9 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87808-49-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD03840737
InChI Key DJTUYAFJAGLQCK-UHFFFAOYSA-N
Synonym methyl2-bromo-4-methylbenzoate, 2-bromo-4-methylbenzoic acid methyl ester, benzoic acid, 2-bromo-4-methyl-, methyl ester, pubchem14035, acmc-209qq7, ksc495m0h, methyl 2-bromo-4-methyl-benzoate, 2-bromo-4-methyl-benzoic acid methyl ester
PubChem CID 21779704
IUPAC Name methyl 2-bromo-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)Br

Specifications

Quantity 250 mg
Formula Weight 229.07
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-bromo-4-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.