Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromo-5-chlorobenzoate 98.0+%, TCI America™
SDP

Catalog No. M203325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M203325G 25 g
M20335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M203325G Supplier TCI America Supplier No. M203325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27007-53-0
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD00144763
InChI Key BIECSXCXIXHDBC-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzoic acid methyl ester, benzoic acid, 2-bromo-5-chloro-, methyl ester, pubchem11238, acmc-1cr8r, ksc494q3f, methyl-2-bromo-5-chlorobenzoate, methyl 2-bromo-5-chloro-benzoate, methyl 2-bromanyl-5-chloranyl-benzoate, 2-bomo-5-chlorobenzoic acid methyl ester
PubChem CID 280500
IUPAC Name methyl 2-bromo-5-chlorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)Br

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 25 g
Formula Weight 249.49
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Methyl 2-Bromo-5-chlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.