Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromohexanoate 98.0+%, TCI America™
SDP

Catalog No. M2378250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2378250G 250 g
M237825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2378250G Supplier TCI America Supplier No. M2378250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5445-19-2
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00045038
InChI Key YGLPDRIMFIXNBI-UHFFFAOYSA-N
Synonym 2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
PubChem CID 95578
IUPAC Name methyl 2-bromohexanoate
SMILES CCCCC(C(=O)OC)Br

Specifications

Color Colorless
Boiling Point 94°C
Quantity 250 g
Formula Weight 209.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromohexanoate
EINECSNumber (2)-3662
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.