Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-(Bromomethyl)-3-nitrobenzoate 98.0+%, TCI America™
SDP

Catalog No. M24221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24221G 1 g
M24225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24221G Supplier TCI America Supplier No. M24221G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98475-07-1
Molecular Formula C9H8BrNO4
Molecular Weight (g/mol) 274.07
MDL Number MFCD04114315
InChI Key FCGIVHSBEKGQMZ-UHFFFAOYSA-N
Synonym methyl 2-bromomethyl-3-nitrobenzoate, 2-bromomethyl-3-nitrobenzoic acid methyl ester, methyl 2-bromomethyl-3-nitro-benzoate, benzoic acid, 2-bromomethyl-3-nitro-, methyl ester, methyl 2-bromomethyl-3-nitrobenzenecarboxylate, 2-bromomethyl-3-nitro benzoic acid methyl ester, methyl2-bromomethyl-3-nitrobenzoate, zlchem 474, pubchem19169
PubChem CID 1487795
IUPAC Name methyl 2-(bromomethyl)-3-nitrobenzoate
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])CBr

Specifications

Melting Point 73°C
Color Yellow
Quantity 1 g
UN Number 3261
Formula Weight 274.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(Bromomethyl)-3-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.