Learn More
Methyl 2-bromothiazole-4-carboxylate, 96%, Thermo Scientific Chemicals
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Quantity | 250 mg |
| Synonym | methyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid methyl ester, 2-bromo-4-methoxycarbonyl-1,3-thiazole, methyl2-bromothiazole-4-carboxylate, methyl-2-bromothiazole-4-carboxylate, 4-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-4-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-4-carboxylic acid methyl ester, pubchem8920, acmc-1c5qt |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 222.06 |
| Percent Purity | 96% |
| Physical Form | Solid |
| Chemical Name or Material | Methyl 2-bromothiazole-4-carboxylate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.