Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (2-Chlorophenyl)acetate 98.0+%, TCI America™
SDP

Catalog No. C1740100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1740100G 100 g
C174025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1740100G Supplier TCI America Supplier No. C1740100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57486-68-7
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00137445
InChI Key KHBWTRFWQROKJZ-UHFFFAOYSA-N
Synonym methyl 2-chlorophenylacetate, methyl 2-2-chlorophenyl acetate, methyl 2-chlorophenyl acetate, 2-chlorophenyl acetic acid methyl ester, benzeneacetic acid, 2-chloro-, methyl ester, 2-chlorophenylacetic acid methylester, 2-chlorobenzeneacetic acid methyl ester, acmc-209lyj, methyl2-chlorophenylacetate
PubChem CID 93683
IUPAC Name methyl 2-(2-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 100 g
Physical Form Clear Liquid at 20°C
Boiling Point 128°C
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl (2-Chlorophenyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.