Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-(Cyanomethyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. C155225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C155225G 25 g
C15525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C155225G Supplier TCI America Supplier No. C155225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4-6-5597
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD01631304
InChI Key CMADSXVGXNLKHA-UHFFFAOYSA-N
Synonym 2-(Cyanomethyl)benzoic Acid Methyl Ester
PubChem CID 588805
IUPAC Name methyl 2-(cyanomethyl)benzoate
SMILES COC(=O)C1=CC=CC=C1CC#N

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 133°C
Quantity 25 g
UN Number 3439
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(Cyanomethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.