Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-methylbutyrate, 98%

Catalog No. AC450230050
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450230050 5 kg
AC450230010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC450230050 Supplier Thermo Scientific Chemicals Supplier No. 450230050
This item is not returnable. View return policy

CAS: 868-57-5 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-57-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009335
InChI Key OCWLYWIFNDCWRZ-UHFFFAOYNA-N
Synonym methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
PubChem CID 13357
IUPAC Name methyl 2-methylbutanoate
SMILES CCC(C)C(=O)OC

Specifications

Density 0.8850g/mL
Boiling Point 115.0°C
Flash Point 18°C
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula CH3CH2CH(CH3)COOCH3
Quantity 5 kg
Specific Gravity 0.885
Solubility Information Solubility in water: practically insoluble
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Methyl 2-methylbutyrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
EINECSNumber 212-778-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.