Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Oxo-1-pyrrolidineacetate 97.0+%, TCI America™
SDP

Catalog No. O020525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O020525G 25 g
O02055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O020525G Supplier TCI America Supplier No. O020525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59776-88-4
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00003198
InChI Key HPDZYDTXCOFUOY-UHFFFAOYSA-N
Synonym 2-Oxo-1-pyrrolidineacetic Acid Methyl Ester, 2-Pyrrolidinone-1-acetic Acid Methyl Ester
PubChem CID 108835
IUPAC Name methyl 2-(2-oxopyrrolidin-1-yl)acetate
SMILES COC(=O)CN1CCCC1=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 157.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Oxo-1-pyrrolidineacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.