Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Oxocyclopentanecarboxylate 97.0+%, TCI America™
SDP

Catalog No. C0812100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0812100ML 100 mL
C081225ML 25 mL
C0812500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0812100ML Supplier TCI America Supplier No. C0812100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10472-24-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00001411
InChI Key PZBBESSUKAHBHD-UHFFFAOYSA-N
Synonym methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane
PubChem CID 66328
IUPAC Name methyl 2-oxocyclopentane-1-carboxylate
SMILES COC(=O)C1CCCC1=O

Specifications

Color Yellow
Boiling Point 111°C
Quantity 100 mL
Formula Weight 142.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Oxocyclopentanecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.