Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate 98.0+%, TCI America™
SDP

Catalog No. M279725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M279725G 25 g
M27975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M279725G Supplier TCI America Supplier No. M279725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57113-90-3
Molecular Formula C13H16N2O6
Molecular Weight (g/mol) 296.279
MDL Number MFCD06797690
InChI Key VEDIIGMQOAWKGH-UHFFFAOYSA-N
Synonym 2-(tert-Butoxycarbonylamino)-3-nitrobenzoic Acid Methyl Ester, Methyl 2-(Boc-amino)-3-nitrobenzoate, 2-(Boc-amino)-3-nitrobenzoic Acid Methyl Ester
PubChem CID 9965968
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-nitrobenzoate
SMILES CC(C)(C)OC(=O)NC1=C(C=CC=C1[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 106°C
Color Yellow
Quantity 25 g
Formula Weight 296.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.