Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,3,3-Trifluoropropionate 98.0+%, TCI America™
SDP

Catalog No. M27831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27831G 1 g
M27835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27831G Supplier TCI America Supplier No. M27831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18830-44-9
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD02262206
InChI Key PMGBATZKLCISOD-UHFFFAOYSA-N
Synonym 3,3,3-Trifluoropropionic Acid Methyl Ester
PubChem CID 2782392
IUPAC Name methyl 3,3,3-trifluoropropanoate
SMILES COC(=O)CC(F)(F)F

Specifications

Color Colorless
Boiling Point 96°C
Quantity 1 g
UN Number 3272
Formula Weight 142.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3,3,3-Trifluoropropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.