Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,4-Dimethoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M280525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M280525G 25 g
M28055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M280525G Supplier TCI America Supplier No. M280525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2150-38-1
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008430
InChI Key BIGQPYZPEWAPBG-UHFFFAOYSA-N
Synonym methyl veratrate, benzoic acid, 3,4-dimethoxy-, methyl ester, 3,4-dimethoxybenzoic acid methyl ester, veratric acid, methyl ester, methylveratrate, pubchem13009, acmc-1cfay, veratric acid methyl ester, methyl 3,4 dimethoxybenzoate, methyl 3, 4-dimethoxybenzoate
PubChem CID 16522
ChEBI CHEBI:86906
IUPAC Name methyl 3,4-dimethoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C(OC)=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 283°C
Quantity 25 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4-Dimethoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.