Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 3-amino-5-(4-bromophenyl)thiophene-2-carboxylate

Catalog No. JRD07691G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD07691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD07691G Supplier Sigma Aldrich Supplier No. JRD07691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromo-1-methyl-1h-imidazole can be used in the synthesis of the following:
  • N-(4-methoxyphenyl)-1-methyl-1h-imidazol-2-amine via c-n coupling with p-anisidine
  • 1,2-bis[2-methyl-5-(1-methyl-1h-imidazol-2-yl)-3-thienyl]-cyclopentene, a ligand which can form bimetallic platinum(ii) complex with potent cytotoxic activity

Specifications

Molecular Formula C12H10BrNO2S
Linear Formula C12H10BrNO2S
MDL Number MFCD04116396
Quantity 1 g
Molecular Weight (g/mol) 312.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.