Learn More
Methyl 3-bromobenzoate, 98+%
Description
Methyl 3-bromobenzoate is used as a reagent in organic synthesis. It is used in the synthesis of tetrahydro dibenzazocine, which acts as an inhibitor of 17beta-hydroxysteroid dehydrogenase type 3. Further, it is used to prepare 3-tert-butylphosphanyl-benzoic acid methyl ester by reacting with tert-butyltrimethylsilylphosphine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 3-bromobenzoate is used as a reagent in organic synthesis. It is used in the synthesis of tetrahydro dibenzazocine, which acts as an inhibitor of 17beta-hydroxysteroid dehydrogenase type 3. Further, it is used to prepare 3-tert-butylphosphanyl-benzoic acid methyl ester by reacting with tert-butyltrimethylsilylphosphine.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | 30°C to 33°C |
| Boiling Point | 139°C to 140°C (35 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.5575 |
| Quantity | 50 g |
| Beilstein | 1940810 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 215.05 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Methyl 3-bromobenzoate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 210-569-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.