Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Chloro-4-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. M213625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M213625G 25 g
M21365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M213625G Supplier TCI America Supplier No. M213625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56525-63-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00205141
InChI Key KTFQDZCNPGFKAH-UHFFFAOYSA-N
Synonym methyl3-chloro-4-methylbenzoate, methyl-3-chloro-4-methylbenzoate, methyl 3-chloro-p-toluate, 2-chloro-4-methoxycarbonyl toluene, 3-chloro-4-methylbenzoic acid methyl ester, benzoic acid, 3-chloro-4-methyl-, methyl ester, pubchem10935, acmc-209lsv, ksc495i9j, methyl-3-chloro-4-metylbenzoate
PubChem CID 2801405
IUPAC Name methyl 3-chloro-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Cl

Specifications

Color White-Yellow
Boiling Point 107°C
Quantity 25 g
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Chloro-4-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.