Learn More
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Physical Form | Clear Liquid at 20°C |
| Synonym | methyl 2-3-chlorophenyl acetate, methyl 3-chlorophenylacetate, methyl 3-chlorophenyl acetate, phenylacetic acid, 3-chloro, methyl ester, benzeneacetic acid, 3-chloro-, methyl ester, methyl2-3-chlorophenyl acetate, methyl 2-3-chlorophenyl ethanoate, 3-chlorophenylacetic acid methyl ester, 3-chlorophenyl acetic acid methyl ester |
| Formula Weight | 184.62 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | Methyl (3-Chlorophenyl)acetate |
Safety and Handling
| TSCA | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
