Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Cyclopropyl-3-oxopropionate 96.0+%, TCI America™
SDP

Catalog No. M227725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M227725G 25 g
M22775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M227725G Supplier TCI America Supplier No. M227725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32249-35-7
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00274090
InChI Key RIJWDPRXCXJDPK-UHFFFAOYSA-N
Synonym 3-Cyclopropyl-3-oxopropionic Acid Methyl Ester
PubChem CID 10130016
IUPAC Name methyl 3-cyclopropyl-3-oxopropanoate
SMILES COC(=O)CC(=O)C1CC1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 g
Formula Weight 142.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Cyclopropyl-3-oxopropionate
EINECSNumber (3)-4627
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.