Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Furancarboxylate 96.0+%, TCI America™
SDP

Catalog No. F046825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F046825G 25 g
F04685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F046825G Supplier TCI America Supplier No. F046825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13129-23-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD06203671
InChI Key ZKHQSQYLKSSYIP-UHFFFAOYSA-N
Synonym 3-Furoic Acid Methyl Ester, 3-Furancarboxylic Acid Methyl Ester, Methyl 3-Furoate
PubChem CID 14918
IUPAC Name methyl furan-3-carboxylate
SMILES COC(=O)C1=COC=C1

Specifications

Color Yellow
Boiling Point 64°C
Quantity 25 g
UN Number 3272
Formula Weight 126.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Furancarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.