Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Methyl 3-Hydroxydecanoate, 3-Hydroxy Fatty Acid Methyl Ester, 25 mg

Catalog No. 502706047
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6047 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6047 Supplier Abcam Supplier No. AB14402725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxy fatty acid methyl ester with >98% purity for use as a standard in biological systems.

  • Molecular weight: 202.29 Da
  • Purity: >98%
  • Composition: 3-Hydroxy fatty acid methyl ester (100% Methyl 3-hydroxydecanoate)
  • Applications: Standard and biological systems

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule Methyl 3-hydroxydecanoate
Quantity 25 mg
For Use With (Application) Standard, Biological systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.