Learn More
Abcam Methyl 3-Hydroxyhexanoate, 3-Hydroxy Fatty Acid Methyl Ester, >98% Purity, 25 mg
Description
- Molecular Weight: 146.18 Da
- Purity: >98%
- Chemical Class: 3-Hydroxy fatty acid methyl ester
- Application: Standard in methanolysis assay and in biological systems
- Composition: 100% Methyl 3-hydroxyhexanoate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Methyl 3-hydroxyhexanoate |
| Buffer | 100% Methyl 3-hydroxyhexanoate |
| Formulation | 100% Methyl 3-hydroxyhexanoate |
| Quantity | 25 mg |
| For Use With (Application) | Methanolysis assay, biological systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.