Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Hydroxyisoxazole-5-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M24411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24411G 1 g
M24415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24411G Supplier TCI America Supplier No. M24411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10068-07-2
Molecular Formula C5H5NO4
Molecular Weight (g/mol) 143.10
MDL Number MFCD00062870
InChI Key BBFWUUBQSXVHHZ-UHFFFAOYSA-N
Synonym methyl 3-hydroxyisoxazole-5-carboxylate, methyl 3-hydroxy-5-isoxazolecarboxylate, methyl 3-hydroxy-1,2-oxazole-5-carboxylate, 3-hydroxyisoxazole-5-carboxylic acid methyl ester, methyl 3-hydroxyl-5-isoxazolecarboxylate, methyl-3-hydroxy-5-isoxazolecarboxylate, 5-isoxazolecarboxylic acid, 2,3-dihydro-3-oxo-, methyl ester, 3-hydroxy-isoxazole-5-carboxylic acid methyl ester, methyl 3-oxo-2,3-dihydro-1,2-oxazole-5-carboxylate, methyl 3-oxo-2h-1,2-oxazole-5-carboxylate
PubChem CID 2724585
IUPAC Name methyl 3-oxo-2,3-dihydro-1,2-oxazole-5-carboxylate
SMILES COC(=O)C1=CC(=O)NO1

Specifications

Melting Point 164°C
Color Yellow
Quantity 1 g
Formula Weight 143.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Hydroxyisoxazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.