Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Methyl 3-Hydroxyoctanoate, Polyester, >98% Purity, 25 mg

Catalog No. 502706045
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6045 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6045 Supplier Abcam Supplier No. AB14404325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medium-chain-length 3-hydroxyalkanoate polyester derivative with >98% purity.

  • Molecular weight: 174.24 Da
  • Purity: >98%
  • Composition: 100% Methyl 3-hydroxyoctanoate
  • Derivative: 3-Hydroxyoctanoic acid (ab144042)
  • Classification: Polyester; medium-chain-length 3-hydroxyalkanoate
  • Storage temperature: Store at -20°C
  • Shipping temperature: Ambient - can ship with ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Methyl 3-hydroxyoctanoate
Buffer 100% Methyl 3-hydroxyoctanoate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Formulation 100% Methyl 3-hydroxyoctanoate
Quantity 25 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.