Learn More
Abcam Methyl 3-hydroxytetradecanoate, 3-Hydroxy Fatty Acid Methyl Ester, >98% Purity, 25 mg
Description
- Purity: >98%
- Molecular Weight: 258.4 Da
- CAS Number: 55682-83-2
- Storage buffer: Constituents: 100% Methyl 3-hydroxytetradecanoate
- Application: Internal standard
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Methyl 3-hydroxytetradecanoate |
| Shipping Condition | Ambient |
| Buffer | 100% Methyl 3-hydroxytetradecanoate |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Methyl 3-hydroxytetradecanoate |
| Quantity | 25 mg |
| For Use With (Application) | Internal standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.