Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Methoxyisobutyrate 98.0+%, TCI America™
SDP

Catalog No. M119125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M119125ML 25 mL
M1191500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M119125ML Supplier TCI America Supplier No. M119125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3852-11-7
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00671553
InChI Key LYLUAHKXJUQFDG-UHFFFAOYNA-N
Synonym 3-Methoxyisobutyric Acid Methyl Ester, Methyl 3-Methoxy-2-methylpropionate, 3-Methoxy-2-methylpropionic Acid Methyl Ester
PubChem CID 107177
IUPAC Name methyl 3-methoxy-2-methylpropanoate
SMILES CC(COC)C(=O)OC

Specifications

Color Colorless
Boiling Point 148°C
Quantity 25 mL
UN Number 1993
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methoxyisobutyrate
EINECSNumber (2)-1405
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.