Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(Trifluoromethyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. M171825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M171825G 25 g
M17185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M171825G Supplier TCI America Supplier No. M171825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2557-13-3
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
InChI Key QQHNNQCWKYFNAC-UHFFFAOYSA-N
Synonym methyl 3-trifluoromethyl benzoate, 3-carbomethoxybenzotrifluoride, 3-trifluoromethyl benzoic acid methyl ester, methyl3-trifluoromethyl benzoate, methyl m-trifluoromethyl benzoate, 3-trifluoromethyl-benzoic aic methyl ester, mtf-bom, methyl m-trifluoromethylbenzoate, 3-methoxycarbonyl benzotrifluoride
PubChem CID 520213
IUPAC Name methyl 3-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
UN Number 3272
Formula Weight 204.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-(Trifluoromethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.