Learn More
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%
Description
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.
Solubility
It is soluble in water, acetone, benzene, ethyl ether
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 163°C to 166°C |
| Quantity | 25 g |
| Beilstein | 1291458 |
| Sensitivity | Hygroscopic |
| Solubility Information | It is soluble in water,acetone,benzene,ethyl ether |
| Optical Rotation | +110° (c=2 in Chloroform) |
| Formula Weight | 282.29 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside |
Safety and Handling
| EINECSNumber | 221-615-2 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.