Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Acetamido-2-methoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M277625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M277625G 25 g
M27765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M277625G Supplier TCI America Supplier No. M277625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4093-29-2
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00065258
InChI Key OERVVBDWGVOBIS-UHFFFAOYSA-N
Synonym 4-Acetamido-2-methoxybenzoic Acid Methyl Ester, Methyl 4-Acetamido-o-anisate, 4-Acetamido-o-anisic Acid Methyl Ester
PubChem CID 77720
IUPAC Name methyl 4-acetamido-2-methoxybenzoate
SMILES COC(=O)C1=CC=C(NC(C)=O)C=C1OC

Specifications

Melting Point 130°C
Color White
Quantity 25 g
Formula Weight 223.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Acetamido-2-methoxybenzoate
EINECSNumber (3)-1662
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.