Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-acetylbenzoate, 99%

Click to view available options
Quantity:
2g
10g
3609-53-8
C10H10O3
178.187
MFCD00216474
QNTSFZXGLAHYLC-UHFFFAOYSA-N
benzoic acid, 4-acetyl-, methyl ester,unii-3s3d53j4nt,4-acetylbenzoic acid methyl ester,methyl4-acetylbenzoate,4-acetyl-benzoic acid methyl ester,methyl 4-acetyl benzoate,pubchem22273,acmc-209ikx,#,ksc497k9b
137990
methyl 4-acetylbenzoate
CC(=O)C1=CC=C(C=C1)C(=O)OC
3609-53-8
C10H10O3
178.187
MFCD00216474
QNTSFZXGLAHYLC-UHFFFAOYSA-N
benzoic acid, 4-acetyl-, methyl ester,unii-3s3d53j4nt,4-acetylbenzoic acid methyl ester,methyl4-acetylbenzoate,4-acetyl-benzoic acid methyl ester,methyl 4-acetyl benzoate,pubchem22273,acmc-209ikx,#,ksc497k9b
137990
methyl 4-acetylbenzoate
CC(=O)C1=CC=C(C=C1)C(=O)OC
Melting Point 92°C to 95°C
Quantity 10g
Formula Weight 178.19
Percent Purity 99%
Chemical Name or Material Methyl 4-acetylbenzoate