Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-acetylbenzoate, 99%

Catalog No. AAB2162604
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2162604 2 g
AAB2162609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2162604 Supplier Thermo Scientific Chemicals Supplier No. B2162604
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3609-53-8 | C10H10O3 | 178.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3609-53-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00216474
InChI Key QNTSFZXGLAHYLC-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl-, methyl ester, unii-3s3d53j4nt, 4-acetylbenzoic acid methyl ester, methyl4-acetylbenzoate, 4-acetyl-benzoic acid methyl ester, methyl 4-acetyl benzoate, pubchem22273, acmc-209ikx, #, ksc497k9b
PubChem CID 137990
IUPAC Name methyl 4-acetylbenzoate
SMILES CC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 92°C to 95°C
Quantity 2 g
Formula Weight 178.19
Percent Purity 99%
Chemical Name or Material Methyl 4-acetylbenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.