Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-aminobutyrate hydrochloride, 99%

Catalog No. AC442910250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC442910250 25 g Glass Bottle
AC442911000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442910250 Supplier Thermo Scientific Chemicals Supplier No. 442910250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13031-60-2 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-60-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00043270
InChI Key WPGPRLVPWACBHW-UHFFFAOYSA-N
Synonym methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
PubChem CID 2756383
IUPAC Name methyl 4-aminobutanoate;hydrochloride
SMILES [Cl-].COC(=O)CCC[NH3+]

Specifications

Melting Point 118°C to 119°C
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Glass Bottle
Quantity 25 g
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material Methyl 4-aminobutyrate hydrochloride
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.