Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Benzyloxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M218025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M218025G 25 g
M21805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M218025G Supplier TCI America Supplier No. M218025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32122-11-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00017613
InChI Key ZLLQTDQOTFCCDF-UHFFFAOYSA-N
PubChem CID 299810
IUPAC Name methyl 4-(benzyloxy)benzoate
SMILES COC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 99°C
Color White
Quantity 25 g
Synonym methyl 4-benzyloxybenzoate, methyl 4-benzyloxy benzoate, 4-benzyloxybenzoic acid methyl ester, methyl4-benzyloxybenzoate, methyl 4-benzyloxy benzenecarboxylate, methyl 4-phenylmethoxy benzoate, methyl-4-benzyloxybenzoate, acmc-209hr7, 4-benzyloxy-benzoic acid methyl ester, benzoic acid,4-phenylmethoxy-, methyl ester
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Benzyloxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.