Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Bromo-2-chlorobenzoate 98.0+%, TCI America™
SDP

Catalog No. M28835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28835G 5 g
M28831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28835G Supplier TCI America Supplier No. M28835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 185312-82-7
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD10566823
InChI Key JSNXIWYWUKTWAX-UHFFFAOYSA-N
Synonym 4-Bromo-2-chlorobenzoic Acid Methyl Ester
PubChem CID 22058730
IUPAC Name methyl 4-bromo-2-chlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Br)Cl

Specifications

Quantity 5 g
Formula Weight 249.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Bromo-2-chlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.