Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Chloro-3-nitrobenzoate 98.0+%, TCI America™
SDP

Catalog No. M177925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M177925G 25 g
M17795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M177925G Supplier TCI America Supplier No. M177925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14719-83-6
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00016989
InChI Key XRTKWPWDSUNLHS-UHFFFAOYSA-N
Synonym methyl 4-chloro-3-nitrobenzate, 4-chloro-3-nitrobenzoic acid methyl ester, methyl-4-chloro-3-nitrobenzoate, benzoic acid, 4-chloro-3-nitro-, methyl ester, acmc-209cya, methyl-3-nitro-4-chlorobenzoate, methyl 3-nitro-4-chlorobenzoate, xrtkwpwdsunlhs-uhfffaoysa, methyl 4-chloro-3-nitro-benzoate, methyl4-chloro-3-nitrobenzoate
PubChem CID 735797
IUPAC Name methyl 4-chloro-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 82°C
Quantity 25 g
Formula Weight 215.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Chloro-3-nitrobenzoate
EINECSNumber (3)-1519
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.