Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (4-Chlorophenyl)acetate 98.0+%, TCI America™
SDP

Catalog No. M203925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M203925G 25 g
M20395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M203925G Supplier TCI America Supplier No. M203925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52449-43-1
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00032743
InChI Key WWIYGBWRUXQDND-UHFFFAOYSA-N
Synonym methyl 4-chlorophenylacetate, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, 4-chloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, 4-chlorophenylacetic acid methyl ester, methyl4-chlorophenylacetate, zlchem 382
PubChem CID 104196
IUPAC Name methyl 2-(4-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 115°C
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl (4-Chlorophenyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.