Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Fluoro-3-hydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M191925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M191925G 25 g
M19195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M191925G Supplier TCI America Supplier No. M191925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 214822-96-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD06797457
InChI Key CUGWNEOTLGLGDG-UHFFFAOYSA-N
PubChem CID 12016892
IUPAC Name methyl 4-fluoro-3-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C=C1)F)O

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Synonym 4-fluoro-3-hydroxybenzoic acid methyl ester, benzoic acid, 4-fluoro-3-hydroxy-, methyl ester, pubchem3567, acmc-209fky, methyl4-fluoro-3-hydroxybenzoate, methyl 4-fluoranyl-3-oxidanyl-benzoate, 3-hydroxy-4-fluorobenzoic acid methyl ester, 4-fluoro-3-hydroxy-benzoic acid methyl ester, benzoic acid,4-fluoro-3-hydroxy-, methyl ester
Formula Weight 170.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Fluoro-3-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.