Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Hydroxy-3,5-dimethoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M280625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M280625G 25 g
M28065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M280625G Supplier TCI America Supplier No. M280625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 884-35-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00017199
InChI Key ZMXJAEGJWHJMGX-UHFFFAOYSA-N
Synonym methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976
PubChem CID 70164
ChEBI CHEBI:45820
IUPAC Name methyl 4-hydroxy-3,5-dimethoxybenzoate
SMILES COC1=CC(=CC(=C1O)OC)C(=O)OC

Specifications

Melting Point 108°C
Color White to Yellow
Quantity 25 g
Formula Weight 212.2
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Hydroxy-3,5-dimethoxybenzoate
EINECSNumber (3)-4633
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.