Learn More
Methyl 4-hydroxybenzoate, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Methyl 4-hydroxybenzoate is used as an anti-fungal agent. It is also used as a preservative in foods, beverages and cosmetics. It acts as an inhibitor of growth of molds and to lesser extent bacteria and as a vehicle for ophthalmic solution.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 125°C to 128°C |
| pH | 5.8 |
| Boiling Point | ∼280°C (decomposition) |
| Odor | Odorless |
| Quantity | 1000 g |
| Beilstein | 509801 |
| Merck Index | 14,6107 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 152.15 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| EINECSNumber | 202-785-7 |
| RTECSNumber | DH2450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.