Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (4-Hydroxybenzoyl)acetate 93.0+%, TCI America™
SDP

Catalog No. H08281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08281G 1 g
H08285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08281G Supplier TCI America Supplier No. H08281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32066-29-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00191516
InChI Key VZOFEVHSVUBEPH-UHFFFAOYSA-N
Synonym (4-Hydroxybenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester
PubChem CID 508465
IUPAC Name methyl 3-(4-hydroxyphenyl)-3-oxopropanoate
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 194.19
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (4-Hydroxybenzoyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.