Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Iodo-3-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. I060125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I060125G 25 g
I06015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I060125G Supplier TCI America Supplier No. I060125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5471-81-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD02683863
InChI Key HCSGWQGKCVQIRM-UHFFFAOYSA-N
Synonym 4-iodo-3-methylbenzoic acid methyl ester, methyl 4-iodo-3-methyl-benzoate, 4-iodo-m-toluic acid methyl ester, benzoic acid, 4-iodo-3-methyl-, methyl ester, methyl4-iodo-3-methylbenzoate, acmc-1anqa, methyl 4-iodo-m-toluate, methyl 3methyl-4-iodobenzoate, methyl 3-methyl-4-iodobenzoate, 4-iodo-m-toluic acid, methyl ester
PubChem CID 231757
IUPAC Name methyl 4-iodo-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)I

Specifications

Melting Point 61°C
Boiling Point 106°C
Quantity 25 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Iodo-3-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.