Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Methylcinnamate 98.0+%, TCI America™
SDP

Catalog No. M28251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28251G 1 g
M28255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28251G Supplier TCI America Supplier No. M28251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7560-43-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00460746
InChI Key WLJBRXRCJNSDHT-UHFFFAOYSA-N
Synonym 4-Methylcinnamic Acid Methyl Ester
PubChem CID 589472
IUPAC Name methyl 3-(4-methylphenyl)prop-2-enoate
SMILES CC1=CC=C(C=C1)C=CC(=O)OC

Specifications

Melting Point 58°C
Color White
Boiling Point 165°C
Quantity 1 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Methylcinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.