Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Methylsalicylate 98.0+%, TCI America™
SDP

Catalog No. M158425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M158425G 25 g
M15845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M158425G Supplier TCI America Supplier No. M158425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4670-56-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00020130
InChI Key UITFCFWKYAOJEJ-UHFFFAOYSA-N
Synonym methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate
PubChem CID 78400
IUPAC Name methyl 2-hydroxy-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)O

Specifications

Color White
Boiling Point 242°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Methyl 4-Methylsalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.