Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-nitro-1-(p-toluenesulfonyl)pyrrole-2-carboxylate, 95%, Thermo Scientific™

Catalog No. AAH3346903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3346903 1 g
AAH33469MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3346903 Supplier Thermo Scientific Chemicals Supplier No. H3346903

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930111-86-7
Molecular Formula C13H12N2O6S
Molecular Weight (g/mol) 324.31
MDL Number MFCD09743415
InChI Key AYFBDJWRDWDJJZ-UHFFFAOYSA-N
Synonym methyl 1-4-methylbenzenesulfonyl-4-nitropyrrole-2-carboxylate, methyl 4-nitro-1-tosyl-1h-pyrrole-2-carboxylate, methyl 4-nitro-1-p-toluenesulfonyl pyrrole-2-carboxylate, methyl 1-4-methylphenyl sulfonyl-4-nitropyrrole-2-carboxylate, methyl 4-nitro-1-toluene-4-sulphonyl-1h-pyrrole-2-carboxylate, methyl 1-4-methylbenzene-1-sulfonyl-4-nitro-1h-pyrrole-2-carboxylate, methyl 4-nitro-1-4-methylphenyl sulphonyl-1h-pyrrole-2-carboxylate
PubChem CID 44784923
IUPAC Name methyl 1-(4-methylphenyl)sulfonyl-4-nitropyrrole-2-carboxylate
SMILES COC(=O)C1=CC(=CN1S(=O)(=O)C1=CC=C(C)C=C1)[N+]([O-])=O

Specifications

Melting Point 198°C to 200°C
Quantity 1 g
Formula Weight 260.25
Percent Purity 95%
Chemical Name or Material Methyl 4-nitro-1-(p-toluenesulfonyl)pyrrole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.